missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA146001
This item is not returnable.
View return policy
Description
This antibody shares 85% sequence homology with rat. Suggested positive control: Hela whole cell extract, antigen standard for CDR2 (transient overexpression lysate).
Using anti-Yo antibodies, three genes have been cloned (CDR 1-3). CDR2 suppresses the transcriptional activity and DNA binding of nuclear factor-kappa B by an unknown mechanism. The neuron specific expression of CDR2 is regulated by a post-transcriptional mechanism.
Specifications
| CDR2 | |
| Polyclonal | |
| Unconjugated | |
| CDR2 | |
| AA617262; CDR2; CDR62; cerebellar degeneration related protein 2; cerebellar degeneration-related 2; cerebellar degeneration-related protein 2; cerebellar degeneration-related protein 2, 62kDa; major Yo paraneoplastic antigen; MGC128861 protein; paraneoplastic cerebellar degeneration-associated antigen; PCD17; RGD1310578; Yo; Yo paraneoplastic antigen | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1039, 12585, 534647 | |
| 4°C, do not freeze | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Phosphate, tris citrate with 0.1% sodium azide; pH 7-8 | |
| P97817, Q01850 | |
| CDR2 | |
| A synthetic peptide made to a C-terminal region of human CDR2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey, Bovine | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction