missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585546
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (88%), Rat (90%), Rhesus Monkey (97%), Bovine (90%). Positive Control: U87-MG, SK-N-SH, IMR32, SK-N-AS. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Using anti-Yo antibodies, three genes have been cloned (CDR 1-3). CDR2 suppresses the transcriptional activity and DNA binding of nuclear factor-kappa B by an unknown mechanism. The neuron specific expression of CDR2 is regulated by a post-transcriptional mechanism.
Specifications
| CDR2 | |
| Polyclonal | |
| Unconjugated | |
| CDR2 | |
| AA617262; CDR2; CDR62; cerebellar degeneration related protein 2; cerebellar degeneration-related 2; cerebellar degeneration-related protein 2; cerebellar degeneration-related protein 2, 62kDa; major Yo paraneoplastic antigen; MGC128861 protein; paraneoplastic cerebellar degeneration-associated antigen; PCD17; RGD1310578; Yo; Yo paraneoplastic antigen | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1039 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q01850 | |
| CDR2 | |
| Recombinant protein encompassing a sequence within the center region of human CDR2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction