missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDR2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-84562-25ul
This item is not returnable.
View return policy
Description
CDR2 Polyclonal antibody specifically detects CDR2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CDR2 | |
| Polyclonal | |
| Western Blot 0.04 - 0.4 ug/mL, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000-1:2500 | |
| CDR62, cerebellar degeneration-related protein (62kD), cerebellar degeneration-related protein 2, cerebellar degeneration-related protein 2, 62kDa, Major Yo paraneoplastic antigen, Paraneoplastic cerebellar degeneration-associated antigen, PCD17, Yo, Yo paraneoplastic antigen | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: GVEKLVPDSLYVPFKEPSQSLLEEMFLTVPESHRKPLKRSSSETILSSLAGSDIVKGHEETCIRRAKAVKQRGISLLHEVDTQYSALKVKYEELLKKCQEEQDSLSHKAVQTSRAAAKDLTGV | |
| 25 μL | |
| Cancer, Immune System Diseases, Immunology, Neuroscience | |
| 1039 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction