missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDR2 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-58345CL1
This item is not returnable.
View return policy
Description
CDR2 Polyclonal antibody specifically detects CDR2 in Bovine,Human,Mouse,Primate samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| CDR2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Bovine, Human, Mouse, Primate | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| CDR62, cerebellar degeneration-related protein (62kD), cerebellar degeneration-related protein 2, cerebellar degeneration-related protein 2, 62kDa, Major Yo paraneoplastic antigen, Paraneoplastic cerebellar degeneration-associated antigen, PCD17, Yo, Yo paraneoplastic antigen | |
| A synthetic peptide made to a C-terminal region of human CDR2 [Swiss-Prot# Q01850] | |
| 0.1 mL | |
| Cancer, Hypoxia, Immune System Diseases, Immunology, Neuroscience, Signal Transduction | |
| 1039 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction