missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDP-Star™ Substrate (0.4 mM Ready-to-Use) with Enhancer

Product Code. 10429145
Change view
Click to view available options
Description:
Emerald-II™ Enhancer
Sapphire-II™ Enhancer
Unit Size:
100mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
10429145 Emerald-II™ Enhancer 100mL
12654335 Sapphire-II™ Enhancer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10429145 Supplier Invitrogen™ Supplier No. T2216
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Ready-to-use chemiluminescent alkaline phosphatase substrate, for protein or nucleic acid blotting on nitrocellulose membranes or solution-based assays.

Chemiluminescent substrates provide highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP). Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.

The CDP-Star™ Substrate with Sapphire-II™ or Emerald-II™ Enhancer is a chemiluminescent alkaline phosphatase substrate, ready-to-use for protein or nucleic acid blotting on nitrocellulose membranes. This substrate can also be used in solution-based assays.

This CDP-Star™ chemiluminescent substrate for alkaline phosphatase let you detect alkaline phosphatase and alkaline phosphatase-labeled molecules with unparalleled sensitivity, speed, and ease.

• Chemiluminescent substrates provide highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP).
• Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.

This versatile chemiluminescent substrate exhibits high sensitivity in membrane-based applications such as Southern, Northern, and Western blotting. This substrate can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays. Maximum light levels are reached in approximately 10 minutes and glow emission persists for several hours. Solution assays require chemiluminescent enhancer. Membrane-based assays may also require it for increased light output and sensitivity.

For Research Use Only. Not for use in diagnostics procedures.

TRUSTED_SUSTAINABILITY

Specifications

Description Emerald-II™ Enhancer
Product Type ELISA Substrate with Enhancer
Quantity 100 mL
Detection Method Chemiluminescence
Form Liquid
Format Bottle
Product Line CDP-Star, NovaBright
Substrate Type Alkaline Phosphatase
Content And Storage Store the product at 4°C.
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.