missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDO1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA538005

Product Code. 15594931

  • £430.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

A recommended positive control is rat liver tissue lysate.

CDO1 (cysteine dioxygenase, type I) belongs to the cysteine dioxygenase family and is involved in organosulfur biosynthesis. CDO1 functions as a dioxygenase that uses iron and zinc as cofactors to catalyze the conversion of L-cysteine and oxygen to 3-sulfinoalanine. CDO1 exists as a monomer and is expressed at high levels in liver and placenta and at lower levels in brain, pancreas and heart. CDO1 is involved in pyruvate-, sulfate- and taurine-related metabolic pathways and is a crucial regulator of cysteine concentrations within the cell.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CDO1
Polyclonal
Unconjugated
Cdo1
1300002L19Rik; Cdo; CDO1; CDOI; CDO-I; cysteine dioxygenase 1, cytosolic; cysteine dioxygenase type 1; Cysteine dioxygenase type I; cysteine dioxygenase, type I; cytosolic cysteine dioxygenase 1; D18Ucla3
Rabbit
Antigen Affinity Chromatography
RUO
1036, 12583, 81718
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
P21816, P60334, Q16878
Cdo1
19 amino acid peptide near the amino terminus of human CDO1.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.