missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDO Rabbit anti-Human, Clone: 76, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | CDO |
|---|---|
| Clone | 76 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Kvantitet och tillgänglighet | |||||
|
18253406
|
Novus Biologicals
NBP2-90033-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18224077
|
Novus Biologicals
NBP2-90033-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
CDO Monoclonal specifically detects CDO in Human samples. It is validated for ELISA.Specifikationer
| CDO | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 50937 | |
| IgG | |
| Protein A purified |
| 76 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Cdon homolog (mouse), CDOORCAM, cell adhesion molecule-related/down-regulated by oncogenes, MGC111524, surface glycoprotein, Ig superfamily member | |
| Recombinant Human CDO protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel