missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDO Antibody (076), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90033F
This item is not returnable.
View return policy
Description
CDO Monoclonal specifically detects CDO in Human samples. It is validated for ELISA.
Specifications
| CDO | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 50937 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 76 | |
| ELISA | |
| Cdon homolog (mouse), CDOORCAM, cell adhesion molecule-related/down-regulated by oncogenes, MGC111524, surface glycoprotein, Ig superfamily member | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CDO (AAI14436.1; Met1-Asp963). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction