missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDNF Monoclonal Antibody (D2), Invitrogen™

Mouse Monoclonal Antibody
£178.00 - £386.00
Specifications
| Antigen | CDNF |
|---|---|
| Clone | D2 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17674346
|
Invitrogen™
MA543893 |
20 μL |
£178.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17664346
![]() |
Invitrogen™
MA543892 |
100 μL |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The conserved dopamine neurotrophic factor (CDNF) is a neurotrophic factor for dopaminergic neurons and highly homologous to the mesencephalic-astrocyte-derived neuro-trophic factor. Somewhat surprisingly, CDCF is expressed at higher levels in tissues such as heart, skeletal muscle and testes than in brain. Similar to the glial cell line-derived neurotrophic factor (GDNF), CDNF can prevent the 6-hydroxylamine (6 OHDA) induced degeneration of dopaminergic neurons in a rat model of Parkinsons disease. Furthermore, CDNF was able to restore the dopaminergic function and prevent the degeneration of dopaminergic neurons in the substantia nigra, suggesting that CDNF might be suitable for the treatment of Parkinsons disease. At least two isoforms of CDNF are known to exist.Specifications
| CDNF | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| 9330140G23; arginine-rich protein mutated in early stage tumors-like 1; arginine-rich, mutated in early stage tumors-like 1; ARMETL1; ARMET-like protein 1; Cdnf; Cerebral dopamine neurotrophic factor; conserved dopamine neurotrophic factor | |
| Cdnf | |
| IgG2b κ | |
| Protein A/G | |
| Antibody |
| D2 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q49AH0 | |
| 441549 | |
| Pro33-Leu187 (Accession Q49AH0), with N-terminal His and GST Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Cdnf |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title