missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CDNF Monoclonal specifically detects CDNF in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | CDNF |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 104 |
| Conjugate | DyLight 550 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | arginine-rich, mutated in early stage tumors-like 1, ARMETL1, ARMET-like protein 1, cerebral dopamine neurotrophic factor, Conserved dopamine neurotrophic factor |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human CDNF (NP_001025125.2; Met1-Leu187). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?