missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDNF Antibody (091), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90638AF350
This item is not returnable.
View return policy
Description
CDNF Monoclonal specifically detects CDNF in Mouse samples. It is validated for ELISA.
Specifications
| CDNF | |
| Monoclonal | |
| Alexa Fluor 350 | |
| arginine-rich, mutated in early stage tumors-like 1, ARMETL1, ARMET-like protein 1, cerebral dopamine neurotrophic factor, Conserved dopamine neurotrophic factor | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CDNF (NP_808315.1; Met 1-Leu 187). | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 441549 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 91 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction