missing translation for 'onlineSavingsMsg'
Learn More

CDKN3, p Thr14 Antibody (4C12), Novus Biologicals™

Product Code. 18378593 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18378593 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18378593 Supplier Novus Biologicals Supplier No. NBP315205

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CDKN3 Monoclonal antibody specifically detects CDKN3 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDKN3
Applications Western Blot, ELISA
Classification Monoclonal
Clone 4C12
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA
Formulation PBS (pH 7.4), 150mM NaCl, and 50% glycerol.
Gene Alias CDI1KAP1, CDK2-associated dual specificity phosphatase, CDK2-associated dual-specificity phosphatase, Cdk-associated protein phosphatase, CIP2, cyclin-dependent kinase inhibitor 3, cyclin-dependent kinase interacting protein 2, Cyclin-dependent kinase interactor 1, Cyclin-dependent kinase-interacting protein 2, EC 3.1.3.16, EC 3.1.3.48, FLJ25787, KAPMGC70625, Kinase-associated phosphatase
Host Species Rabbit
Immunogen A synthesized peptide derived from human Phospho-CDKN3 (T14). UniProt ID (P06493/P24941/Q00526).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Protein Phosphatase, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1033
Target Species Human
Content And Storage Store at -20 to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.