missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDKN3 Antibody (OTI2E11), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70399PEATT594
This item is not returnable.
View return policy
Description
CDKN3 Monoclonal specifically detects CDKN3 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| CDKN3 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| CDKN3 | |
| Full length recombinant protein of human CDKN3 (NP_005183) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG1 |
| Flow Cytometry | |
| OTI2E11 | |
| Flow Cytometry | |
| CDI1KAP1, CDK2-associated dual specificity phosphatase, CDK2-associated dual-specificity phosphatase, Cdk-associated protein phosphatase, CIP2, cyclin-dependent kinase inhibitor 3, cyclin-dependent kinase interacting protein 2, Cyclin-dependent kinase interactor 1, Cyclin-dependent kinase-interacting protein 2, EC 3.1.3.16, EC 3.1.3.48, FLJ25787, KAPMGC70625, Kinase-associated phosphatase | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication, Core ESC Like Genes, Protein Phosphatase, Stem Cell Markers | |
| 1033.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction