missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDKN2B Polyclonal Antibody

CDKN2B Polyclonal Antibody
Brand: Invitrogen PA599491
This item is not returnable.
View return policy
Description
CDKN2B Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
| CDKN2B | |
| Polyclonal | |
| Unconjugated | |
| CDKN2B | |
| AV083695; CDK inhibitory protein; CDK4B inhibitor; CDK4I; Cdkn2b; cyclin dependent kinase inhibitor 2B; Cyclin dependent kinase inhibitor 2B (p15, inhibits CDK4); cyclin-dependent kinase 4 inhibitor B; cyclin-dependent kinase inhibitor 2B (p15, inhibits CDK4); cyclin-dependent kinase inhibitor p15; cyclin-dependent kinase inhibitor p15INK4b; cyclin-dependent kinase inhibitor protein; cyclin-dependent kinases 4 and 6 binding protein; Ink4; INK4B; MTS2; MTS-2; Multiple tumor suppressor 2; p14_CDK inhibitor; p14_INK4B; p14-INK4b; P15; p15 CDK inhibitor; p15(INK4b); p15_INK4B; p15INK4b; p15-INK4b; TP15 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1030, 12579, 25164 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P42772, P55271, P55272 | |
| CDKN2B | |
| A synthesized peptide derived from human CDKN2B(Accession P42772), corresponding to amino acid residues L93-D138. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction