missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CDKN1B/CEN12p FISH Probe
Click to view available options
Quantity:
100 μL
200 μL
Unit Size:
100µL
200µL
Description
Includes
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Specifications
Specifications
| Content And Storage | Store at 4°C in the dark. |
| Description | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
| Format | Tube |
| For Use With (Application) | FISH-Ce |
| Gene ID (Entrez) | 1027 |
| Includes | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
| Quantity | 200 μL |
| Target | Human |
| Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction