Learn More
CDKL5 Antibody (CL4888) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61146APC
Product Code. 30287823
  • £558.00 / 0.10mL
Description
CDKL5 Monoclonal antibody specifically detects CDKL5 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CDKL5
Monoclonal
APC
cyclin-dependent kinase-like 5, serine/threonine kinase 9, serine/threonine-protein kinase 9
This antibody was developed using a recombinant protein derived from O76039, with the exact immunogen sequence remaining proprietary.
0.1 mL
Protein Kinase
6792
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL4888
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified