missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDKL4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15732112 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15732112 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15732112 Supplier Invitrogen™ Supplier No. PA564347

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: FHGISIPEPE DMETLEEKFS DVHPVALNFM KGCLKMNPDD RLTCSQLLES SYFDS Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 80%.

CDKL4 (Cyclin Dependent Kinase Like 4) is a Protein Coding gene. Diseases associated with CDKL4 include Orofaciodigital Syndrome Vi. Gene Ontology (GO) annotations related to this gene include transferase activity, transferring phosphorus-containing groups and protein tyrosine kinase activity. An important paralog of this gene is CDKL1. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDKL4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDKL4
Gene Accession No. Q5MAI5
Gene Alias AU067824; CDKL4; cyclin dependent kinase like 4; cyclin-dependent kinase-like 4; EC 2.7.11.22; Gm942
Gene Symbols CDKL4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDKL4. Recombinant protein control fragment (Product #RP-103662).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 344387
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.