missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDKL4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550928
This item is not returnable.
View return policy
Description
CDKL4 Polyclonal Antibody detects endogenous levels of CDKL4 protein.
CDKL4 (Cyclin Dependent Kinase Like 4) is a Protein Coding gene. Diseases associated with CDKL4 include Orofaciodigital Syndrome Vi. Gene Ontology (GO) annotations related to this gene include transferase activity, transferring phosphorus-containing groups and protein tyrosine kinase activity. An important paralog of this gene is CDKL1. [GeneCards].
Specifications
| CDKL4 | |
| Polyclonal | |
| Unconjugated | |
| CDKL4 | |
| AU067824; CDKL4; cyclin dependent kinase like 4; cyclin-dependent kinase-like 4; EC 2.7.11.22; Gm942 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 344387 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5MAI5 | |
| CDKL4 | |
| Synthesized peptide derived from the C-terminal region of human CdkL4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction