missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CDKL4 Polyclonal specifically detects CDKL4 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | CDKL4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | cyclin-dependent kinase-like 4, EC 2.7.11, EC 2.7.11.22 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human CDKL4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?