missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDKL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545998
This item is not returnable.
View return policy
Description
Peptide sequence: EKYEKIGKIG EGSYGVVFKC RNRDTGQIVA IKKFLESEDD PVIKKIALRE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
This gene product is a member of a large family of CDC2-related serine/threonine protein kinases. It accumulates primarily in the nucleus. Alternative splice variants encoding different protein isoforms have been described, but their full-length nature has not been determined.
Specifications
| CDKL1 | |
| Polyclonal | |
| Unconjugated | |
| CDKL1 | |
| 4933411O17Rik; CDC2-related kinase 1; Cdkl1; cyclin dependent kinase like 1; cyclin-dependent kinase-like 1; cyclin-dependent kinase-like 1 (CDC2-related kinase); KKIALRE; P42; protein kinase p42 KKIALRE; serine/threonine protein kinase KKIALRE; serine/threonine-protein kinase KKIALRE; zgc:101002 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 8814 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q00532 | |
| CDKL1 | |
| Synthetic peptide directed towards the N-terminal of human CDKL1 (aa 3-52). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction