missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDKL1 Antibody (OTI1D9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46217
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
CDKL1 Monoclonal specifically detects CDKL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| CDKL1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry 1:150 | |
| CDC2-related kinase 1, cyclin-dependent kinase-like 1, cyclin-dependent kinase-like 1 (CDC2-related kinase), EC 2.7.11, EC 2.7.11.22, KKIALRE, P42, Protein kinase p42 KKIALRE, serine/threonine protein kinase KKIALRE, Serine/threonine-protein kinase KKIALRE | |
| Mouse | |
| 41.7 kDa | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 8814 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1D9 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| CDKL1 | |
| Human recombinant protein fragment corresponding to amino acids 152-357 of human CDKL1(NP_004187) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion