missing translation for 'onlineSavingsMsg'
Learn More

CDKL1 Antibody (2H6), Novus Biologicals™

Product Code. 18387398 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18387398 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18387398 Supplier Novus Biologicals Supplier No. H00008814M05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CDKL1 Monoclonal antibody specifically detects CDKL1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDKL1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2H6
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_004187
Gene Alias CDC2-related kinase 1, cyclin-dependent kinase-like 1, cyclin-dependent kinase-like 1 (CDC2-related kinase), EC 2.7.11, EC 2.7.11.22, KKIALRE, P42, Protein kinase p42 KKIALRE, serine/threonine protein kinase KKIALRE, Serine/threonine-protein kinase KKIALRE
Host Species Mouse
Immunogen CDKL1 (NP_004187, 259 a.a. ~ 358 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. YPALGLLKGCLHMDPTQRLTCEQLLHHPYFENIREIEDLAKEHNKPTRKTLRKSRKHHCFTETSKLQYLPQLTGSSILPALDNKKYYCDTKKLNYRFPNI
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 8814
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.