missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDKAL1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22988-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody recognizes the 65 kDa isoform of CDKAL1.
The protein encoded by this gene is a member of the methylthiotransferase family. The function of this gene is not known. Genome-wide association studies have linked single nucleotide polymorphisms in an intron of this gene with susceptibility to type 2 diabetes.Specifications
| CDKAL1 | |
| Polyclonal | |
| Unconjugated | |
| CDKAL1 | |
| CDKAL1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54901, 68916 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5VV42, Q91WE6 | |
| CDKAL1 | |
| CDKAL1 Fusion Protein Ag19060 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction