missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDKAL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112318
This item is not returnable.
View return policy
Description
CDKAL1 Polyclonal Antibody for Western Blot, IP
The protein encoded by this gene is a member of the methylthiotransferase family. The function of this gene is not known. Genome-wide association studies have linked single nucleotide polymorphisms in an intron of this gene with susceptibility to type 2 diabetes.
Specifications
| CDKAL1 | |
| Polyclonal | |
| Unconjugated | |
| Cdkal1 | |
| 1190005B03Rik; 6620401C13Rik; CDK5 regulatory subunit associated protein 1 like 1; CDK5 regulatory subunit associated protein 1-like 1; CDK5 regulatory subunit-associated protein 1-like 1; CDKAL; CDKAL1; Threonylcarbamoyladenosine tRNA methylthiotransferase; tRNA-t(6)A37 methylthiotransferase | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 54901 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q5VV42 | |
| Cdkal1 | |
| E. coli-derived Human CDKAL1 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction