missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDKAL1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142330
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
The protein encoded by this gene is a member of the methylthiotransferase family. The function of this gene is not known. Genome-wide association studies have linked single nucleotide polymorphisms in an intron of this gene with susceptibility to type 2 diabetes.
Specifications
| CDKAL1 | |
| Polyclonal | |
| Unconjugated | |
| Cdkal1 | |
| 1190005B03Rik; 6620401C13Rik; CDK5 regulatory subunit associated protein 1 like 1; CDK5 regulatory subunit associated protein 1-like 1; CDK5 regulatory subunit-associated protein 1-like 1; CDKAL; CDKAL1; Threonylcarbamoyladenosine tRNA methylthiotransferase; tRNA-t(6)A37 methylthiotransferase | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 54901 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q5VV42 | |
| Cdkal1 | |
| Peptide with sequence C-QVKKQRTKDLSR. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction