missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDKAL1 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98668AF405
This item is not returnable.
View return policy
Description
CDKAL1 Polyclonal specifically detects CDKAL1 in Human samples. It is validated for Western Blot.
Specifications
| CDKAL1 | |
| Polyclonal | |
| Western Blot | |
| CDK5 regulatory subunit associated protein 1-like 1, CDK5 regulatory subunit-associated protein 1-like 1, FLJ20342, FLJ46705, MGC75469 | |
| Produced in rabbits immunized with E. coli-derived Human CDKAL1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 54901 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction