missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11705-1-AP-20UL
This item is not returnable.
View return policy
Description
CDK9 (cyclin-dependent kinase 9) is a member of the cyclin-dependent protein kinase family. It is involved in the regulation of transcription. CDK9 is a member of the cyclin-dependent kinase pair (CDK9/cyclin-T) complex. This complex facilitates the transition from abortive to productive elongation by phosphorylating the CTD of the large subunit of RNA polymerase II, POLR2A, SUPT5H, and RDBP. The complex is inactive when in the 7SK snRNP complex form. CDK9 phosphorylates EP300, MYOD1, RPB1/POLR2A, AR, DSIF, and NELF. It regulates cytokine inducible transcription networks by facilitating promoter recognition of target transcription factors. CDK9 promotes RNA synthesis in genetic programs for cell growth, differentiation, and viral pathogenesis. The CDK9/cyclin-K complex also has kinase activity towards CTD of RNAPII and can substitute for CDK9/cyclin-T P-TEFb in vitro. Chronic activation of CDK9 causes cardic myocyte enlargement leading to cardiac hypertrophy and confers a predisposition to heart failure.Specifications
| CDK9 | |
| Polyclonal | |
| Unconjugated | |
| CDK9 | |
| C 2k, CDC2L4, CDK9, Cell division protein kinase 9, CTK1, cyclin dependent kinase 9, PITALRE, TAK | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1025, 107951, 362110 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P50750, Q641Z4, Q99J95 | |
| Cdk9 | |
| CDK9 Fusion Protein Ag2318 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction