missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK9-IN-2, MedChemExpress
Description
CDK9-IN-2 is a special cyclin-dependent kinase 9 (CDK9) inhibitor, extracted from patent WO/2012131594A1, compound CDKI(8), has an IC50 of 5 nM and 7 nM in H929 multiple myeloma(MM) cell line (72 hours) and A2058 skin cell line (72 hours), respectively.
Specifications
Specifications
| Chemical Name or Material | CDK9-IN-2 |
| CAS | 1263369-28-3 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C23H25ClFN5 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 50 mg/mL (117.39 mM; Need ultrasonic) |
| SMILES | ClC1=CN=C(N[C@@H]2CC[C@@H](N)CC2)C=C1C3=NC(NCC4=CC(F)=CC=C4)=CC=C3 |
| Molecular Weight (g/mol) | 425.93 |
| Formula Weight | 425.93 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?