missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CDK9/Cyclin T1, His Tag Recombinant Protein

Product Code. 11513627
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11513627

Brand: Invitrogen™ PV4131

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length.

Cyclin-dependent kinase 9 (CDK9) and Cyclin T1 form the positive transcription elongation factor b (p-TEFb) complex, which is crucial for transcriptional regulation and mRNA maturation. CDK9 is a transcriptional kinase expressed widely across human tissues, with notably higher levels in terminally differentiated cells, influencing transcriptional elongation by phosphorylating the RNA polymerase II C-terminal domain. This kinase's gene is mapped to chromosome 9q34.1 and produces two isoforms through two distinct promoters. CDK9, when coupled with Cyclin T1, modulates oncogene expression and activity, representing essential roles in cell survival and proliferation. The dysregulation of CDK9/Cyclin T1's activity has connections to tumorigenesis, highlighting their importance in cancer biology as potential therapeutic targets. Their complex operation also involves acetylation changes crucial for functional modulation. The interaction of CDK9 with other regulatory proteins emphasizes its versatile roles in cellular physiology beyond what is seen in classical cell cycle control.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O60563, P50750
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 0.02% Triton X-100, 150mM NaCl, 0.5mM EDTA, 2mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 1025, 904
Molecular Weight (g/mol) 46.9-84.8 kDa
Name Human CDK9/Cyclin T1, His Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias C-2 k; CDC2L4; CDC2-related kinase; Cdk9; Cell division cycle 2-like protein kinase 4; Cell division protein kinase 9; CTK1; cyclin dependent kinase 9; cyclin-dependent kinase 9; cyclin-dependent kinase 9 (CDC2-related kinase); PITALRE; RP11-228B15.5; serine/threonine protein kinase PITALRE; Serine/threonine-protein kinase PITALRE; TAK; Tat-associated kinase complex catalytic subunit
Common Name CDK9/Cyclin T1
Gene Symbol CCNT1, CDK9
KinaseFamily CDK and CHK Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical binding/kinetic, Biochemical ADP accumulation
Kinase Group Ser⁄Thr Kinases: CMCG Group
Shipping Condition Dry Ice
System Compatibility Adapta™
Target Entry CDK9/Cyclin T1
Target Gene cyclin-dependent kinase 9
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Cell Cycle & Proliferation, Signal Transduction
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.