missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Cdk9 Monoclonal antibody specifically detects Cdk9 in Drosophila,Human samples. It is validated for Immunoprecipitation,Western Blot,ELISA,Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | Cdk9 |
| Applications | Immunoprecipitation, Western Blot, ELISA, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Clone | 2D7 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | AAH01968 |
| Gene Alias | C-2k, CDC2L4CTK1, CDC2-related kinase, Cell division cycle 2-like protein kinase 4, Cell division protein kinase 9, cyclin-dependent kinase 9, cyclin-dependent kinase 9 (CDC2-related kinase), EC 2.7.11, EC 2.7.11.22, EC 2.7.11.23, PITALRE, serine/threonine protein kinase PITALRE, Serine/threonine-protein kinase PITALRE, TAK |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?