missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK8, Mouse anti-Human, Clone: 6H5, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00001024-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: QQQGNNHTNGTGHPGNQDSSHTQGPPLKKVRVVPPTTTSGGLIMTSDYQRSNPHAAYPNPGPSTSQPQSSMGYSATSQQPPQYSHQTHRYSpecifications
| CDK8 | |
| Monoclonal | |
| Unconjugated | |
| cyclin-dependent kinase 8 | |
| K35/MGC126074/MGC126075 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
| ELISA, Western Blot | |
| 6H5 | |
| In 1x PBS, pH 7.4 | |
| NM_001260 | |
| CDK8 | |
| CDK8 (NP_001251, 375 a.a. ∼ 464 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 1024 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction