missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CDK8/Cyclin C, His Tag Recombinant Protein

Product Code. p-7075727
Click to view available options
Quantity:
10 μg
100 μg
Unit Size:
100µg
10µg
This item is not returnable. View return policy

Product Code. 15468543

Brand: Invitrogen™ PR7261B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

CDK8 phosphorylates the carboxy-terminal domain of the largest subunit of RNA polymerase II. This protein is coexpressed and copurified with Cyclin C. CDK8 is part of a family of proteins designated as cdks are critical regulators of cell cycle progression. The prototype member of this family, p34/Cdc 2, and a related protein Cdk 2, function late in the cycle while Cdk 4 and Cdk 6 are critically involved in G1 to S progression. Because of analogy with yeast Cdk 8, it has been proposed that the human Cdk 8 Cyclin C may interact with RNA polymerase, thus influencing the transcription mechanism(s) that lead to cell division.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49336
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 0.02% Triton X-100, 2mM DTT, 150mM NaCl, 0.5mM EDTA, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 1024
Molecular Weight (g/mol) 97 kDa
Name Human CDK8/Cyclin C, His Tag
pH Range 7.5
Purification Method Purified
Quantity 100 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Cdk8; CDK8 protein kinase; cell division protein kinase 8; Cell division protein kinase 8-like protein; cyclin dependent kinase 8; cyclin-dependent kinase 8; K35; mediator complex subunit CDK8; Mediator of RNA polymerase II transcription subunit CDK8; MGC126074; MGC126075; protein kinase K35; RGD1560888; wu:fk95h11
Common Name CDK8
Gene Symbol Cdk8
KinaseFamily CDK and CHK Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Kinase Group Ser⁄Thr Kinases: CMCG Group
Shipping Condition Dry Ice
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Research Category Cell Cycle & Proliferation, Signal Transduction
Purity or Quality Grade 80% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.