missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CDK8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29273
This item is not returnable.
View return policy
Description
CDK8 Polyclonal antibody specifically detects CDK8 in Human samples. It is validated for Immunohistochemistry
Specifications
| CDK8 | |
| Polyclonal | |
| Tris-buffered Saline 0.1% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry | |
| Unconjugated | |
| CDK8 protein kinase, Cell division protein kinase 8, cyclin-dependent kinase 8, EC 2.7.11, EC 2.7.11.22, EC 2.7.11.23, K35, Mediator complex subunit CDK8, Mediator of RNA polymerase II transcription subunit CDK8, MGC126074, MGC126075, Protein kinase K35 | |
| Between 420 and 470 | |
| 25 μg | |
| Cell Cycle and Replication | |
| 1024 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction