missing translation for 'onlineSavingsMsg'
Learn More

CDK8 Antibody [DyLight 405], Novus Biologicals™

Product Code. 30735306 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30735306 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735306 Supplier Novus Biologicals Supplier No. AF6809E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

CDK8 Polyclonal antibody specifically detects CDK8 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK8
Applications Immunohistochemistry
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias CDK8 protein kinase, Cell division protein kinase 8, cyclin-dependent kinase 8, EC 2.7.11, EC 2.7.11.22, EC 2.7.11.23, K35, Mediator complex subunit CDK8, Mediator of RNA polymerase II transcription subunit CDK8, MGC126074, MGC126075, Protein kinase K35
Host Species Sheep
Immunogen E. coli-derived recombinant human CDK8, Asn380-Tyr464, Accession # P49336
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 1024
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.