missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CDK7/Cyclin H/MNAT1, His Tag Recombinant Protein
Human CDK7/Cyclin H/MNAT1, His Tag Recombinant Protein
Brand: Invitrogen PV3868
Code : 50
Additional Details : Weight : 0.05250kg
Description
The trimeric complex of CDK7, cyclin H and MNAT1, functions as a Cdk-activating kinase (CAK). It is an essential component of the transcription factor TFIIH, that is involved in transcription initiation and DNA repair. This complex is thought to serve as a direct link between the regulation of transcription and the cell cycle.Specifications
NP_001790, Q130R I NP_001230 I NP_002422 | |
1022, 1385, 1387, 1390, 4331, 902 | |
10μg | |
CDK and CHK Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: CMCG Group | |
Adapta™ | |
cyclin-dependent kinase 7 | |
Kinases & Inhibitors | |
Liquid | |
Cell Cycle & Proliferation, DNA Replication & Repair, Signal Transduction |
Kinase Assay | |
43.2⁄42.6⁄40.5 kDa | |
CDK7/Cyclin H/MNAT1 | |
Human | |
His Tag | |
Biochemical binding/kinetic, Biochemical ADP accumulation | |
Dry Ice | |
CDK7 | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |
For Research Use Only. Not for use in diagnostic procedures.