missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX Cdk7/Cyclin H/MAT1 (CAK complex) Protein, Active

Product Code. 17300224
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17300224

Brand: Eurofins DiscoverX 14476

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active complex of recombinant full length human Cdk7, containing a C-terminal His6-tag, recombinant full-length human Cyclin H untagged and recombinant full-length human MAT1 containing an N-terminal GST-tag, for use in Kinase Assays.

Cyclin-dependent Kinase 7 (cdk7) is a member of the cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This protein forms a trimeric complex with cyclin H and MAT1, which functions as a Cdk-activating kinase (CAK). It is an essential component of the transcription factor TFIIH, that is involved in transcription initiation and DNA repair. This protein is thought to serve as a direct link between the regulation of transcription and the cell cycle.

GenBank X79193 / GenBank U11791 / GenBank X87843

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 39.9 kDa (CDK7), 37.8 kDa (Cyclin H) and 62.8 kDa (MAT1)
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag C-terminal 6His / untagged / N-terminal GST
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Full-length
Purity or Quality Grade Combined ≥54%
Protein CDK7/Cyclin E/MAT1, active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.