missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19117-1-AP-150UL
This item is not returnable.
View return policy
Description
IARS2 belongs to the class-I aminoacyl-tRNA synthetase family. Members of class I have two highly conserved sequence motifs. They aminoacylate at the 2′-OH of an adenosine nucleotide, and are usually monomeric or dimeric (one or two subunits, respectively). Both classes of aminoacyl-tRNA synthetases are multidomain proteins. The catalytic domains of all the aaRSs of a given class are found to be homologous to one another, while class I and class II aaRSs are unrelated to one another. The class I aaRSs have the ubiquitous Rossmann fold and have the antiparallel beta-strands architecture while the class II aaRSs have a unique fold made up of antiparallel beta-strands.Specifications
| CDK6 | |
| Polyclonal | |
| Unconjugated | |
| CDK6 | |
| CDK6, Cell division protein kinase 6, cyclin dependent kinase 6, PLSTIRE | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1021, 114483, 12571 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q00534, Q64261 | |
| Cdk6 | |
| CDK6 Fusion Protein Ag5600 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction