Learn More
Cdk6 Monoclonal Antibody (K6.83 (DCS-83)), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513333
Description
MA5-13333 targets Cdk6 in ICC/IF, immunohistochemistry (frozen), immunohistochemistry (paraffin), immunoprecipitation, and Western blot applications and shows reactivity with Human, mouse, and Rat samples. The MA5-13333 immunogen is purified recombinant cdk6 protein.
Cdk6 Monoclonal antibody specifically detects Cdk6 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.
Specifications
| Cdk6 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| Q00534, Q64261 | |
| CDK6 | |
| Purified recombinant cdk6 protein | |
| 500 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| K6.83 (DCS-83) | |
| Unconjugated | |
| CDK6 | |
| 5830411I20; AI504062; CDK6; CDKN6; Cell division protein kinase 6; CR2 protein kinase; Crk2; cyclin dependent kinase 6; cyclin-dependent kinase 6; MCPH12; MGC59692; PLSTIRE; Serine/threonine-protein kinase PLSTIRE | |
| Mouse | |
| Protein G | |
| RUO | |
| 1021, 114483, 12571 | |
| 4° C | |
| IgG1 |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.