missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK6 Monoclonal Antibody (K6.83 (DCS-83)), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen AHZ0232
This item is not returnable.
View return policy
Description
This antibody recognizes and epitope at aa 30-65.
CDK6 functions as a cell-cycle initiator protein. This protein is coexpressed and copurified with CyclinD1. They are critical regulators of cell cycle progression. The prototype member of this family, p34cdc2, and a related protein cdk2, function late in the cycle while cdk4 and cdk6 are critically involved in G1 to S progression. CDK6 is essential for cell proliferation within the dentate gyrus of the hippocampus and the cuventricular zone of the lateral ventricles. Mutations in the gene results in microcephaly 12, primary, autosomal recessive (MCPH12).
Specifications
| CDK6 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| Q00534, Q64261 | |
| CDK6 | |
| Purified recombinant cdk6 protein. | |
| 500 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| K6.83 (DCS-83) | |
| Unconjugated | |
| CDK6 | |
| 5830411I20; AI504062; CDK6; CDKN6; Cell division protein kinase 6; CR2 protein kinase; Crk2; cyclin dependent kinase 6; cyclin-dependent kinase 6; MCPH12; MGC59692; PLSTIRE; Serine/threonine-protein kinase PLSTIRE | |
| Mouse | |
| Protein G | |
| RUO | |
| 1021, 114483, 12571 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction