Learn More
Eurofins DiscoverX Cdk6/Cyclin D3 Protein, Active
Description
Mammalian cells express two structurally redundant kinases (Cdk4 and Cdk6) and three structurally redundant partner cyclins (D1, D2 and D3), with predominant expressions of Cdk4 and D1 in mesenchymal cells and Cdk6 and D3 in hematopietic cells. Cdk6 is though to be functionally redundant with Cdk4 and to play the same role in hematopoietic cells as does Cdk4 in fibroblasts. Cyclin D3 is widely expressed in a variety of cells and has been suggested to play a role in the G1-S transition of hematopoietic cells and fibroblasts. Furthermore, recent studies have implicated the involvement of this cyclin subtype in a variety of decisions on cell fate, including proliferation, apoptosis and differentiation.
GenBank X66365 / EMBL M90814
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 270 mM sucrose, 150 mM NaCl, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol, 0.1 mM EGTA, 0.03% Brij-35. Frozen solution. |
| Molecular Weight (g/mol) | 38 kDa (CDK6) and 59 kDa (Cyclin D3). |
| Quantity | 250 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His / N-terminal GST |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Full-length |
| Purity or Quality Grade | Combined ≥71% ratio 1:05 to 1:1 CDK6:cyclinD3 |
| Protein | CDK6/Cyclin D3, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.