missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK5RAP3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538872
This item is not returnable.
View return policy
Description
This gene encodes a protein that has been reported to function in signaling pathways governing transcriptional regulation and cell cycle progression. It may play a role in tumorigenesis and metastasis. A pseudogene of this gene is located on the long arm of chromosome 20. Alternative splicing results in multiple transcript variants that encode different isoforms. [provided by RefSeq, May 2013].
Specifications
| CDK5RAP3 | |
| Polyclonal | |
| Unconjugated | |
| CDK5RAP3 | |
| IC53-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80278, 80279, 80280 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96JB5, Q99LM2, Q9JLH7 | |
| CDK5RAP3 | |
| A synthetic peptide derived from the internal region of human CDK5RAP3 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction