missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK5RAP3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£342.00 - £470.00
Specifications
| Antigen | CDK5RAP3 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18494701
|
Novus Biologicals
NBP1-86779-25ul |
25 μL |
£342.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18249856
|
Novus Biologicals
NBP1-86779 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDK5RAP3 Polyclonal specifically detects CDK5RAP3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CDK5RAP3 | |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication | |
| C53, CDK5 activator-binding protein C53, CDK5 regulatory subunit associated protein 3, FLJ13660, HSF-27, IC53CDK5 regulatory subunit-associated protein 3, ischemic heart CDK5 activator-binding protein C53, LXXLL/leucine-zipper-containing ARFbinding protein, LZAP, MST016, OK/SW-cl.114, Protein HSF-27 | |
| CDK5RAP3 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 80279 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ITGENVRGELLALVKDLPSQLAEIGAAAQQSLGEAIDVYQASVGFVCESPTEQVLPMLRFVQKRGNSTVYEWRTGTEPSVVERPHLEELPEQV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title