missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK5RAP3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92612-0.1ml
This item is not returnable.
View return policy
Description
CDK5RAP3 Polyclonal antibody specifically detects CDK5RAP3 in Human samples. It is validated for Western Blot
Specifications
| CDK5RAP3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| C53, CDK5 activator-binding protein C53, CDK5 regulatory subunit associated protein 3, FLJ13660, HSF-27, IC53CDK5 regulatory subunit-associated protein 3, ischemic heart CDK5 activator-binding protein C53, LXXLL/leucine-zipper-containing ARFbinding protein, LZAP, MST016, OK/SW-cl.114, Protein HSF-27 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human CDK5RAP3 (NP_788276.1). MEDHQHVPIDIQTSKLLDWLVDRRHCSLKWQSLVLTIREKINAAIQDMPESEEIAQLLSGSYIHYFHCLRILDLLKGTEASTKNIFGRYSSQRMKDWQEIIALYEKDNTYLVELSSLLVRNVNYEIPSLKKQIAKCQQLQQEYSRKEEECQAGAAEMREQFYHSCKQYGITGENVRGELLALVKDLPSQLAEIGAAAQQSLGEAIDVYQASVGFVCESPTEQVLPMLRFVQKRGNSTVYEWRTGTEPSVVERPHLEELPEQVAEDAIDWGDFGVEAVSEGTDSGISAEAAGIDWGIFPES | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 80279 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction