missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDK5RAP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116623
This item is not returnable.
View return policy
Description
Positive Samples: LO2, HeLa Immunogen sequence: GQRLLAEMDI QTQEAPSSTS QELGTKGPHP APLSKFVSSV STAKLTLEEA YRRLKLLWRV SLPEDGQCPL HCEQIGEMKA EVTKLHKKLF EQEKKLQNTM KLLQLSKRQE KVIFDQLVVT HKILRKARGN LELRPGGAHP GTCSPSRPGS.
CDK5RAP2 is a regulator of CDK5 (cyclin-dependent kinase 5) activity. The protein is localized to the centrosome and Golgi complex and interacts with CDK5R1 and pericentrin (PCNT). CDK5RAP2 plays a role in centriole engagement and microtubule nucleation, and has been linked to primary microcephaly and Alzheimer's disease. Alternative splicing results in multiple transcript variants.
Specifications
| CDK5RAP2 | |
| Polyclonal | |
| Unconjugated | |
| CDK5RAP2 | |
| C48; CDK5 activator-binding protein C48; CDK5 regulatory subunit associated protein 2; CDK5 regulatory subunit-associated protein 2; CDK5RAP2; centrosomal protein 215 kDa; Centrosome-associated protein 215; centrosomin; Cep215; KIAA1633; MCPH3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55755 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.25 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q96SN8 | |
| CDK5RAP2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1744-1893 of human CDK5RAP2 (NP_0607194). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction