Learn More
Invitrogen™ CDK5RAP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583824
Description
Immunogen sequence: KLHNQEQVIK HLTESTNQKD VLLQKFNEKD LEVIQQNCYL MAAEDLELRS EGLITEKCSS QQPPGSKTIF SKEKKQSSDY EELIQVLKKE QDIYTHLVKS LQESDSINNL QAELNKIFAL RKQLEQDVLS YQNLRKTLEE QISEIRRR Highest antigen sequence indentity to the following orthologs: Rat - 66%, Mouse - 66%.
CDK5RAP2 is a regulator of CDK5 (cyclin-dependent kinase 5) activity. The protein is localized to the centrosome and Golgi complex and interacts with CDK5R1 and pericentrin (PCNT). CDK5RAP2 plays a role in centriole engagement and microtubule nucleation, and has been linked to primary microcephaly and Alzheimer's disease. Alternative splicing results in multiple transcript variants.
Specifications
| CDK5RAP2 | |
| Polyclonal | |
| Unconjugated | |
| CDK5RAP2 | |
| C48; CDK5 activator-binding protein C48; CDK5 regulatory subunit associated protein 2; CDK5 regulatory subunit-associated protein 2; CDK5RAP2; centrosomal protein 215 kDa; Centrosome-associated protein 215; centrosomin; Cep215; KIAA1633; MCPH3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55755 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96SN8 | |
| CDK5RAP2 | |
| Recombinant protein corresponding to Human CDK5RAP2. Recombinant protein control fragment (Product #RP-106126). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.