Learn More
CDK5RAP2 Antibody (CL3392) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59027APC
Product Code. 30301732
  • £558.00 / 0.10mL
Description
CDK5RAP2 Monoclonal antibody specifically detects CDK5RAP2 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CDK5RAP2
Monoclonal
APC
C48, CDK5 activator-binding protein C48, CDK5 regulatory subunit associated protein 2, CDK5 regulatory subunit-associated protein 2, centrosomal protein 215 kDa, Centrosome-associated protein 215, Cep215, DKFZp686B1070, DKFZp686D1070, FLJ10867, KIAA1633microcephaly, primary autosomal recessive 3, MCPH3
This antibody was developed using a recombinant protein derived from Q96SN8, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Cycle and Replication, Cellular Markers, Chromatin Research
55755
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL3392
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified