missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDK5RAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144840
This item is not returnable.
View return policy
Description
Immunogen sequence: RLKDDVPEEV KLRRLEELIT IFREEATKAN QTSVGCTQLV LVEGLSKRSA TDLCGRNDGN LKVIFPDAEM EDVNNPGLRV RAQPGDYVLV KITSAS Antigen sequence identity: Mouse 89%; Rat 86%.
Neuronal CDC2-like kinase, which is involved in the regulation of neuronal differentiation, is composed of a catalytic subunit, CDK5, and an activating subunit, p25NCK5A. The protein encoded by this gene binds to p25NCK5A and therefore may be involved in neuronal differentiation. Multiple transcript variants exist for this gene, but the full-length natures of only two have been determined.
Specifications
| CDK5RAP1 | |
| Polyclonal | |
| Unconjugated | |
| CDK5RAP1 | |
| 2310066P17Rik; C20orf34; C42; CDK5 activator-binding protein C42; CDK5 regulatory subunit associated protein 1; CDK5 regulatory subunit-associated protein 1; CDK5 regulatory subunit-associated protein 1-like protein; CDK5RAP1; CDK5RAP1.3; CDK5RAP1.4; CGI-05; HSPC167; mitochondrial tRNA methylthiotransferase CDK5RAP1; CDK5 regulatory subunit-associated protein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51654 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96SZ6 | |
| CDK5RAP1 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 485-580. Recombinant protein control fragment (Product #RP-105389). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction