missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK5RAP1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92870-0.1ml
This item is not returnable.
View return policy
Description
CDK5RAP1 Polyclonal antibody specifically detects CDK5RAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CDK5RAP1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:100, Immunohistochemistry-Paraffin | |
| C20orf34, C42, CDK5 activator-binding protein C42, CDK5 regulatory subunit associated protein 1, CDK5 regulatory subunit-associated protein 1, CDK5RAP1.3, CDK5RAP1.4, CGI-05, chromosome 20 open reading frame 34, HSPC167 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-350 of human CDK5RAP1 (NP_057492.2). MHPLQCVLQVQRSLGWGPLASVSWLSLRMCRAHSSLSSTMCPSPERQEDGARKDFSSRLAAGPTFQHFLKSASAPQEKLSSEVEDPPPYLMMDELLGRQRKVYLETYGCQMNVNDTEIAWSILQKSGYLRTSNLQEADVILLVTCSIREKAEQTIWNRLHQLKALKTRRPRSRVPLRIGILGCMAERLKEEILNREKMVDILAGPDAYRDLPRLLAVAESGQQAANVLLSLDETYADVMPVQTSASATSAFVSIMRGCDNMCSYCIVPFTRGRERSRPIASILEEVKKLSEQGLKEVTLLGQNVNSFRDNSEVQFNSAVPTNLSRGFTTNYKTKQGGLRFAHLLDQVSRV | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 51654 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction