missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK5R2/p39 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27058-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a neuron-specific activator of CDK5 kinase. It associates with CDK5 to form an active kinase. This protein and neuron-specific CDK5 activator CDK5R1/p39NCK5A both share limited similarity to cyclins, and thus may define a distinct family of cyclin-dependent kinase activating proteins.Specifications
| CDK5R2/p39 | |
| Polyclonal | |
| Unconjugated | |
| CDK5R2 | |
| CDK5 activator 2, CDK5R2, NCK5AI, P39, p39I, p39nck5ai | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12570, 501164, 8941 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O35926, Q13319 | |
| Cdk5r2 | |
| CDK5R2/p39 Fusion Protein Ag25834 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction