Learn More
CDK5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10430-1-AP-150UL
 View more versions of this product
Product Code. 16876813
  • £349.00 / 150µL
Description
Cyclin-dependent kinase-5 (CDK5) is a member of the cyclin-dependent kinase family of serine/threonine kinases. Its mRNA and protein are expressed in the kidneys, testes, and ovaries. And Its activity has been detected almost exclusively in brain extracts. Similar to other Cdks, monomeric Cdk5 displays no enzymatic activity; however, Cdk5 is not activated bycyclins. Instead, Cdk5 activity requires association with one of two brain-specific regulatory subunits, called p35 and p39. These two activators regulate the spatial and temporal expression of active Cdk5, restricting its activity primarily to post-mitotic neurons.
Specifications
Specifications
CDK5
Polyclonal
Unconjugated
CDK5
CDK5, Cell division protein kinase 5, cyclin dependent kinase 5, PSSALRE, TPKII catalytic subunit
Rabbit
Antigen Affinity Chromatography
RUO
1020, 12568, 140908
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P49615, Q00535, Q03114
Cdk5
CDK5 Fusion Protein Ag0686
150 μL
Primary
Human, Mouse, Rat